ENA184121345 Enamine

2-oxo-2,3-dihydro-1H-indole-5-sulfonamide

  • MDL number MFCD05844462
  • Cas Number : 175075-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41204
Availability details for
Quantity
  • Availability
  • Add to Cart