ENA184121353 Enamine

(4-aminophenyl)urea

  • MDL number MFCD09047111
  • Cas Number : 21492-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-164
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27704
Availability details for
Quantity
  • Availability
  • Add to Cart