ENA184121375 Enamine

5-(chloromethyl)-7-hydroxypyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD01451972
  • Cas Number : 925675-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 369...371
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41254
Availability details for
Quantity
  • Availability
  • Add to Cart