ENA184121386 Enamine

1,2,3,4-tetrahydroquinoline-6-carboxamide

  • MDL number MFCD09803679
  • Cas Number : 1016689-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41269
Availability details for
Quantity
  • Availability
  • Add to Cart