ENA184121394 Enamine

3-acetamido-4-methylbenzoic acid

  • MDL number MFCD00020381
  • Cas Number : 6946-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41286
Availability details for
Quantity
  • Availability
  • Add to Cart