ENA184121395 Enamine

N-[4-(aminomethyl)phenyl]-3-[benzyl(methyl)amino]propanamide

  • MDL number MFCD09809995
  • Cas Number : 1016683-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41288
Availability details for
Quantity
  • Availability
  • Add to Cart