ENA184121400 Enamine

3-chloro-N-(4-chlorophenyl)propanamide

  • MDL number MFCD02286134
  • Cas Number : 19314-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40914
Availability details for
Quantity
  • Availability
  • Add to Cart