ENA184121403 Enamine

2-(2-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline

  • MDL number MFCD02720592
  • Cas Number : 380196-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40916

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40916
Availability details for
Quantity
  • Availability
  • Add to Cart