ENA184121406 Enamine

3-amino-N,N,4-trimethylbenzamide

  • MDL number MFCD09736902
  • Cas Number : 76765-68-9
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41315
Availability details for
Quantity
  • Availability
  • Add to Cart