ENA184121417 Enamine

1-[(2,4-difluorophenyl)carbamoyl]piperidine-3-carboxylic acid

  • MDL number MFCD08444836
  • Cas Number : 923207-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41337
Availability details for
Quantity
  • Availability
  • Add to Cart