ENA184121420 Enamine

2-[1-(2,4-difluorophenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD09049199
  • Cas Number : 926239-71-6
  • Molecular Weight : 266.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(2,4-difluorophenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid
Inchi Code 1S/C13H12F2N2O2/c1-7-10(6-13(18)19)8(2)17(16-7)12-4-3-9(14)5-11(12)15/h3-5H,6H2,1-2H3,(H,18,19)
Inchi Key OOEMUJWRASPUTQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41340

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-41340
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart