ENA184121430 Enamine

2-(2,6-dimethylphenoxy)ethanamine hydrochloride

  • MDL number MFCD01106697
  • Cas Number : 313527-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41355
Availability details for
Quantity
  • Availability
  • Add to Cart