ENA184121433 Enamine

N-[(3-fluoro-4-methylphenyl)methyl]piperidine-4-carboxamide

  • MDL number MFCD08443493
  • Cas Number : 926272-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41360
Availability details for
Quantity
  • Availability
  • Add to Cart