ENA184121436 Enamine

7-tert-butyl[1,2,4]triazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD07186563
  • Cas Number : 883546-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41364

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41364
Availability details for
Quantity
  • Availability
  • Add to Cart