ENA184121438 Enamine

4-(2-chlorophenyl)-3-methyl-1,2-oxazol-5-amine

  • MDL number MFCD08446441
  • Cas Number : 924871-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41367
Availability details for
Quantity
  • Availability
  • Add to Cart