ENA184121440 Enamine

1'-(butan-2-yl)-3',5'-dimethyl-1H,1'H-[3,4'-bipyrazole]-5-carboxylic acid

  • MDL number MFCD08741873
  • Cas Number : 1048925-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41369
Availability details for
Quantity
  • Availability
  • Add to Cart