ENA184121442 Enamine

3-(3,4,5-trimethyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD06011102
  • Cas Number : 890593-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41371
Availability details for
Quantity
  • Availability
  • Add to Cart