ENA184121444 Enamine

methyl 2,4-dioxo-4-(1,3,5-trimethyl-1H-pyrazol-4-yl)butanoate

  • MDL number MFCD05861678
  • Cas Number : 890624-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41373
Availability details for
Quantity
  • Availability
  • Add to Cart