ENA184121445 Enamine

[5-chloro-1-(4-fluorophenyl)-3-methyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD08443512
  • Cas Number : 926210-77-7
  • Molecular Weight : 240.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40948
Availability details for
Quantity
  • Availability
  • Add to Cart