ENA184121456 Enamine

2-(3,4-dichlorophenyl)-2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}ethan-1-amine

  • MDL number MFCD11618148
  • Cas Number : 1041546-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41386
Availability details for
Quantity
  • Availability
  • Add to Cart