ENA184121471 Enamine

3-(1-butyl-3,5-dimethyl-1H-pyrazol-4-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD07186519
  • Cas Number : 890624-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41418
Availability details for
Quantity
  • Availability
  • Add to Cart