ENA184121478 Enamine

3-tert-butyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00462163
  • Cas Number : 83405-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41426
Availability details for
Quantity
  • Availability
  • Add to Cart