ENA184121479 Enamine

1,3-dimethyl-4-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD03063405
  • Cas Number : 3654-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41427

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-41427
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart