ENA184121485 Enamine

4-(cyclopentyloxy)benzene-1-sulfonyl chloride

  • MDL number MFCD08687521
  • Cas Number : 914203-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41433
Availability details for
Quantity
  • Availability
  • Add to Cart