ENA184121494 Enamine

3-(1H-1,3-benzodiazol-1-yl)-2-methylpropanoic acid

  • MDL number MFCD02737575
  • Cas Number : 58555-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41456
Availability details for
Quantity
  • Availability
  • Add to Cart