ENA184121498 Enamine

3-methyl-4-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD00831955
  • Cas Number : 60419-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41461

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-41461
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart