ENA184121508 Enamine

6-cyclopropyl-3-propyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09728608
  • Cas Number : 953741-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40973
Availability details for
Quantity
  • Availability
  • Add to Cart