ENA184121510 Enamine

2-(1,3-benzodioxol-5-ylmethoxy)isonicotinonitrile

  • MDL number MFCD09932918
  • Cas Number : 869299-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41483
Availability details for
Quantity
  • Availability
  • Add to Cart