ENA184121511 Enamine

2-(difluoromethoxy)-5-methoxyaniline

  • MDL number MFCD09049113
  • Cas Number : 926238-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41485
Availability details for
Quantity
  • Availability
  • Add to Cart