ENA184121514 Enamine

[(2-chloroethyl)sulfinyl]benzene

  • MDL number MFCD00040896
  • Cas Number : 27998-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34...36
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41515
Availability details for
Quantity
  • Availability
  • Add to Cart