ENA184121535 Enamine

ethyl (allylamino)acetate

  • MDL number MFCD00599116
  • Cas Number : 3182-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40992
Availability details for
Quantity
  • Availability
  • Add to Cart