ENA184121543 Enamine

1-[2-(phenylsulfanyl)ethyl]piperazine dihydrochloride

  • MDL number MFCD11839747
  • Cas Number : 1172282-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41001
Availability details for
Quantity
  • Availability
  • Add to Cart