ENA184121549 Enamine

1-[(2E)-3-thien-2-ylprop-2-enoyl]piperazine

  • MDL number MFCD08444704
  • Cas Number : 58955-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41011
Availability details for
Quantity
  • Availability
  • Add to Cart