ENA185118451 Enamine

trimethyl(3-{[(propan-2-ylimino)methylidene]amino}propyl)azanium iodide

  • MDL number MFCD11857822
  • Cas Number : 856357-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41492

Safety Information

msds https://www.enaminestore.com/catalog/EN300-41492
Availability details for
Quantity
  • Availability
  • Add to Cart