ENA185118453 Enamine

4-[(3-methylbutoxy)methyl]piperidine hydrochloride

  • MDL number MFCD11857825
  • Cas Number : 1171432-92-0
  • Molecular Weight : 221.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41544
Availability details for
Quantity
  • Availability
  • Add to Cart