ENA185118454 Enamine

6-methylquinoline-5-sulfonyl chloride

  • MDL number MFCD11857827
  • Cas Number : 1118787-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41566

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-41566
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart