ENA185118462 Enamine

2-[4-(3-aminopropyl)-3-methyl-1H-pyrazol-1-yl]ethan-1-ol

  • MDL number MFCD11857847
  • Cas Number : 1170583-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41809
Availability details for
Quantity
  • Availability
  • Add to Cart