ENA185118476 Enamine

3-(piperazine-1-carbonyl)-1H-indole hydrochloride

  • MDL number MFCD11857864
  • Cas Number : 1172808-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41988
Availability details for
Quantity
  • Availability
  • Add to Cart