ENA185118499 Enamine

3-amino-N,4-dimethylbenzene-1-sulfonamide

  • MDL number MFCD09881074
  • Cas Number : 1017450-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41479
Availability details for
Quantity
  • Availability
  • Add to Cart