ENA185118503 Enamine

3-hydroxy-4-(2-hydroxyethyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD07636873
  • Cas Number : 42562-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41497
Availability details for
Quantity
  • Availability
  • Add to Cart