ENA185118508 Enamine

3-{6,8-dimethyl-[1,2,4]triazolo[4,3-b]pyridazin-7-yl}propanoic acid

  • MDL number MFCD09881042
  • Cas Number : 1000339-20-7
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-41502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41502
Availability details for
Quantity
  • Availability
  • Add to Cart