ENA185118510 Enamine

6-(4-chlorophenyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD06653399
  • Cas Number : 147269-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41504
Availability details for
Quantity
  • Availability
  • Add to Cart