ENA185118518 Enamine

7-hydrazinyl-5-methyl-2-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyrimidine

  • MDL number MFCD06740805
  • Cas Number : 890590-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41513
Availability details for
Quantity
  • Availability
  • Add to Cart