ENA185118520 Enamine

3-(azepan-1-yl)propanoic acid hydrochloride

  • MDL number MFCD11502900
  • Cas Number : 136671-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41516
Availability details for
Quantity
  • Availability
  • Add to Cart