ENA185118527 Enamine

3-[3,5-dimethyl-1-(prop-2-en-1-yl)-1H-pyrazol-4-yl]-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD07186545
  • Cas Number : 890625-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41526
Availability details for
Quantity
  • Availability
  • Add to Cart