ENA185118532 Enamine

5-(2,4-dimethoxyphenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD05170177
  • Cas Number : 879765-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41531
Availability details for
Quantity
  • Availability
  • Add to Cart