ENA185118539 Enamine

7-tert-butyl-2-methyl-[1,2,4]triazolo[1,5-a]pyrimidine-5-carboxylic acid

  • MDL number MFCD07400762
  • Cas Number : 887672-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41538
Availability details for
Quantity
  • Availability
  • Add to Cart