ENA185118544 Enamine

1-(3-aminophenyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD11505648
  • Cas Number : 1155123-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41543
Availability details for
Quantity
  • Availability
  • Add to Cart