ENA185118548 Enamine

N-methyl-2-(piperidin-4-yl)-N-(propan-2-yl)acetamide hydrochloride

  • MDL number MFCD11815777
  • Cas Number : 1171318-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41547
Availability details for
Quantity
  • Availability
  • Add to Cart