ENA185118572 Enamine

2-(3-methyl-4-nitrophenyl)-1,3,4-oxadiazole

  • MDL number MFCD11833085
  • Cas Number : 1118786-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-33668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33668
Availability details for
Quantity
  • Availability
  • Add to Cart